Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063921.7 | NP_496322.2 | 5 - 6 | 2381 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |