Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129177.4 | NP_001122649.1 | 10 - 11 | 1380 | 79 | |
NM_001267395.3 | NP_001254324.1 | 6 - 7 | 742 | 79 | ||
NM_064144.6 | NP_496545.1 | 10 - 11 | 1395 | 79 | ||
GenBank mRNA | L39212.1 | - | 10 - 11 | 1404 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |