Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027317.8 | NP_001022488.1 | 14 - 15 | 4898 | 77 | |
NM_001267510.3 | NP_001254439.1 | 15 - 16 | 4309 | 77 | ||
NM_001267511.3 | NP_001254440.1 | 13 - 14 | 4528 | 77 | ||
NM_001267517.3 | NP_001254446.1 | 16 - 17 | 5086 | 77 | ||
NM_001350388.3 | NP_001337294.1 | 26 | 7453 | 77 | ||
NM_001377905.1 | NP_001364664.1 | 25 - 26 | 7346 | 77 | ||
NM_064643.10 | NP_497044.3 | 16 - 17 | 5086 | 77 | ||
NM_064645.6 | NP_497046.1 | 16 - 17 | 5177 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |