Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065104.7 | NP_497505.3 | 4 - 5 | 588 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |