Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136364.4 | NP_001129836.1 | 6 - 7 | 557 | 69 | |
NM_065330.7 | NP_497731.1 | 6 - 7 | 486 | 69 | ||
GenBank mRNA | AF083224.1 | - | 6 - 7 | 508 | 69 | |
AY204167.1 | - | 5 - 6 | 487 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |