Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136364.4 | NP_001129836.1 | 8 - 9 | 895 | 86 | |
NM_065330.7 | NP_497731.1 | 8 - 9 | 824 | 86 | ||
GenBank mRNA | AF083224.1 | - | 8 - 9 | 846 | 86 | |
AY204167.1 | - | 7 - 8 | 825 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |