Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027473.4 | NP_001022644.1 | 8 - 9 | 1752 | 78 | |
NM_001027475.3 | NP_001022646.1 | 7 - 8 | 1476 | 78 | ||
GenBank mRNA | AF031838.1 | - | 7 - 8 | 1483 | 78 | |
AF031840.1 | - | 7 - 8 | 1483 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |