Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027391.8 | NP_001022562.1 | 15 - 16 | 2648 | 68 | |
GenBank mRNA | AF256466.1 | - | 15 - 16 | 2638 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |