Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065754.6 | NP_498155.3 | 3 - 4 | 433 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |