Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368430.2 | NP_001355512.1 | 4 - 5 | 616 | 106 | |
NM_001368431.3 | NP_001355513.1 | 4 - 5 | 574 | 106 | ||
NM_001368432.3 | NP_001355514.1 | 3 | 481 | 106 | ||
NM_001368433.3 | NP_001355515.1 | 3 | 481 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |