Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027395.5 | NP_001022566.1 | 11 - 12 | 1917 | 117 | |
NM_001027396.5 | NP_001022567.1 | 11 - 12 | 1719 | 117 | ||
NM_001379772.1 | NP_001367825.1 | 13 - 14 | 2430 | 117 | ||
NM_066066.7 | NP_498467.3 | 13 - 14 | 2393 | 117 | ||
GenBank mRNA | AF399852.1 | - | 10 - 11 | 1697 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |