Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066277.5 | NP_498678.3 | 21 - 22 | 6492 | 75 | |
GenBank mRNA | AF245435.1 | - | 21 - 22 | 6469 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |