Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066502.6 | NP_498903.3 | 13 - 14 | 1435 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |