Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066620.6 | NP_499021.1 | 10 - 11 | 2542 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |