Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027542.4 | NP_001022713.1 | 2 - 3 | 972 | 75 | |
NM_001027543.4 | NP_001022714.1 | 2 - 3 | 978 | 75 | ||
NM_001027544.3 | NP_001022715.1 | 2 - 3 | 971 | 75 | ||
NM_066812.7 | NP_499213.1 | 2 - 3 | 978 | 75 | ||
GenBank mRNA | U73679.1 | - | 2 - 3 | 983 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |