Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sid-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067422.7 | NP_499823.2 | 4 - 5 | 869 | 92 | |
GenBank mRNA | AY466439.1 | - | 4 - 5 | 891 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |