Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307237.2 | NP_001294166.1 | 3 - 4 | 435 | 83 | |
NM_001307238.2 | NP_001294167.1 | 3 - 4 | 586 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |