Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047473.4 | NP_001040938.1 | 6 - 7 | 1227 | 72 | |
NM_001047474.3 | NP_001040939.1 | 4 - 5 | 663 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |