Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ints-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307283.3 | NP_001294212.1 | 7 - 8 | 1683 | 105 | |
NM_001307284.2 | NP_001294213.1 | 7 - 8 | 1355 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |