Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383170.1 | 8 - 9 | 2385 | 150 | ||
NM_069369.5 | NP_501770.1 | 9 - 10 | 1720 | 150 | ||
NM_069370.4 | NP_501771.1 | 8 - 9 | 1450 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |