Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356908.3 | NP_001343633.1 | 2 - 3 | 347 | 83 | |
NM_001372721.2 | NP_001359917.1 | 1 - 2 | 253 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |