Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356908.3 | NP_001343633.1 | 3 | 447 | 89 | |
NM_001372721.2 | NP_001359917.1 | 2 - 3 | 353 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |