Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307367.3 | NP_001294296.1 | 2 - 3 | 443 | 70 | |
NM_068011.4 | NP_500412.1 | 3 - 4 | 764 | 70 | ||
GenBank mRNA | AF201079.1 | - | 3 - 4 | 764 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |