Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068479.1 | NP_500880.1 | 5 - 6 | 665 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |