Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lips-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083237.3 | NP_001076706.1 | 5 - 6 | 658 | 83 | |
NM_001268659.2 | NP_001255588.1 | 5 - 6 | 764 | 83 | ||
NM_001268660.3 | NP_001255589.1 | 5 - 6 | 661 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |