Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129363.4 | NP_001122835.2 | 20 - 21 | 8446 | 98 | |
NM_001268672.3 | NP_001255601.1 | 26 - 27 | 9109 | 98 | ||
NM_001268673.3 | NP_001255602.1 | 27 - 28 | 8872 | 98 | ||
NM_001268674.4 | NP_001255603.2 | 17 - 18 | 7684 | 98 | ||
NM_001268675.3 | NP_001255604.1 | 16 - 17 | 7633 | 98 | ||
NM_001307045.3 | NP_001293974.1 | 29 - 30 | 9754 | 98 | ||
NM_001307046.3 | NP_001293975.1 | 28 - 29 | 9727 | 98 | ||
NM_069872.6 | NP_502273.2 | 26 - 27 | 8972 | 98 | ||
NM_069873.6 | NP_502274.2 | 29 - 30 | 9802 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |