Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129363.4 | NP_001122835.2 | 24 - 25 | 20025 | 104 | |
NM_001268672.3 | NP_001255601.1 | 30 - 31 | 20688 | 104 | ||
NM_001268673.3 | NP_001255602.1 | 31 - 32 | 20451 | 104 | ||
NM_001268674.4 | NP_001255603.2 | 21 - 22 | 19263 | 104 | ||
NM_001268675.3 | NP_001255604.1 | 20 - 21 | 19212 | 104 | ||
NM_001307045.3 | NP_001293974.1 | 33 - 34 | 21333 | 104 | ||
NM_001307046.3 | NP_001293975.1 | 32 - 33 | 21306 | 104 | ||
NM_069872.6 | NP_502273.2 | 30 - 31 | 20551 | 104 | ||
NM_069873.6 | NP_502274.2 | 33 - 34 | 21381 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |