Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268672.3 | NP_001255601.1 | 9 - 10 | 1545 | 72 | |
NM_001268673.3 | NP_001255602.1 | 9 - 10 | 1545 | 72 | ||
NM_001307045.3 | NP_001293974.1 | 9 - 10 | 1545 | 72 | ||
NM_001307046.3 | NP_001293975.1 | 9 - 10 | 1545 | 72 | ||
NM_069872.6 | NP_502273.2 | 9 - 10 | 1672 | 72 | ||
NM_069873.6 | NP_502274.2 | 9 - 10 | 1545 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |