Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069888.9 | NP_502289.2 | 3 - 4 | 483 | 59 | |
GenBank mRNA | DQ140399.1 | - | 3 - 4 | 481 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |