Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268709.3 | NP_001255638.1 | 5 - 6 | 1146 | 64 | |
NM_001268710.3 | NP_001255639.1 | 9 - 10 | 1626 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |