Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070117.9 | NP_502518.2 | 6 - 7 | 1013 | 74 | |
NM_070118.7 | NP_502519.2 | 5 - 6 | 1020 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |