Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070232.6 | NP_502633.1 | 9 - 10 | 1515 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |