Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lst-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276882.3 | NP_001263811.1 | 6 - 7 | 1113 | 90 | |
NM_001307159.3 | NP_001294088.1 | 1 - 2 | 140 | 90 | ||
NM_001383241.1 | 4 - 5 | 782 | 90 | |||
NM_001383242.1 | 5 - 6 | 1423 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |