Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070341.3 | NP_502742.1 | 4 - 5 | 523 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |