Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssu-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268875.3 | NP_001255804.1 | 3 - 4 | 529 | 104 | |
NM_001268876.2 | NP_001255805.1 | 3 - 4 | 534 | 104 | ||
NM_001268877.3 | NP_001255806.1 | 3 - 4 | 612 | 104 | ||
NM_001306937.4 | NP_001293866.1 | 3 - 4 | 730 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |