Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047542.3 | NP_001041007.1 | 8 - 9 | 1446 | 125 | |
NM_001047543.4 | NP_001041008.1 | 5 - 6 | 936 | 125 | ||
NM_001268952.3 | NP_001255881.1 | 6 - 7 | 1227 | 125 | ||
NM_001268953.4 | NP_001255882.1 | 4 - 5 | 741 | 125 | ||
NM_001307194.3 | NP_001294123.1 | 8 - 9 | 1446 | 125 | ||
NM_001307195.3 | NP_001294124.1 | 6 - 7 | 1227 | 125 | ||
NM_001307196.3 | NP_001294125.1 | 5 - 6 | 936 | 125 | ||
NM_001307197.4 | NP_001294126.1 | 4 - 5 | 731 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |