Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070521.2 | NP_502922.1 | 3 - 4 | 604 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |