Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313231.3 | NP_001300160.1 | 1 - 2 | 187 | 82 | |
NM_001313232.3 | NP_001300161.1 | 1 - 2 | 52 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |