Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028790.6 | NP_001023961.1 | 4 - 5 | 441 | 124 | |
NM_001316802.4 | NP_001303731.1 | 4 - 5 | 445 | 124 | ||
NR_133607.1 | 4 - 5 | 441 | 124 | |||
NR_133608.1 | 4 - 5 | 441 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |