Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380620.1 | NP_001367562.1 | 3 - 4 | 648 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |