Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071372.2 | NP_503773.2 | 1 - 2 | 190 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |