Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071489.1 | NP_503890.1 | 3 - 4 | 445 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |