Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hex-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072088.7 | NP_504489.3 | 7 - 8 | 1204 | 133 | |
GenBank mRNA | AM748821.2 | - | 7 - 8 | 1175 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |