Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072150.7 | NP_504551.3 | 2 - 3 | 375 | 100 | |
GenBank mRNA | U09669.1 | - | 2 - 3 | 258 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |