Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083280.3 | NP_001076749.1 | 11 - 12 | 1580 | 81 | |
NM_001373040.1 | NP_001360140.1 | 11 - 12 | 1580 | 81 | ||
NM_001383319.1 | 11 - 12 | 1558 | 81 | |||
GenBank mRNA | AF338767.1 | - | 11 - 12 | 1567 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |