Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028786.7 | NP_001023957.1 | 2 - 3 | 296 | 85 | |
NM_001028787.6 | NP_001023958.1 | 2 - 3 | 299 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |