Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307793.2 | NP_001294722.1 | 5 - 6 | 897 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |