Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073606.5 | NP_506007.1 | 6 - 7 | 719 | 92 | |
NR_001507.1 | 7 - 8 | 698 | 92 | |||
GenBank mRNA | AY557405.1 | - | 6 - 7 | 676 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |