Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073759.3 | NP_506160.1 | 11 - 12 | 1182 | 176 | |
GenBank mRNA | AY008131.1 | - | 7 - 8 | 751 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |