Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nid-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269536.4 | NP_001256465.1 | 8 - 9 | 2571 | 97 | |
NM_001269537.3 | NP_001256466.1 | 8 - 9 | 2577 | 97 | ||
NM_001269538.3 | NP_001256467.1 | 8 - 9 | 2577 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |